Assay Details
| Gene Symbol: EXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193360.1 | NP_001180289.1 | 10 | 3757 | 65 | |
| NM_001193361.1 | NP_001180290.1 | 9 | 3673 | 65 | ||
| NM_001193362.1 | NP_001180291.1 | 10 | 3916 | 65 | ||
| NM_001193363.1 | NP_001180292.1 | 9 | 3832 | 65 | ||
| NM_018199.3 | NP_060669.1 | 8 | 3293 | 65 | ||
| NR_034164.1 | 11 | 3847 | 65 | |||
| NR_034165.1 | 11 | 4006 | 65 | |||
| XM_005267817.3 | XP_005267874.1 | 8 | 3223 | 65 | ||
| XM_011536908.2 | XP_011535210.1 | 8 | 3311 | 65 | ||
| XM_017021421.1 | XP_016876910.1 | 9 | 3356 | 65 | ||
More Information
Set Membership: |
|

EXD2
Genomic Map