Assay Details
| Gene Symbol: SLC39A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252148.1 | NP_001239077.1 | 6 | 2254 | 76 | |
| NM_001252151.1 | NP_001239080.1 | 7 | 1750 | 76 | ||
| NM_001252152.1 | NP_001239081.1 | 8 | 1738 | 76 | ||
| NM_018375.4 | NP_060845.2 | 7 | 2323 | 76 | ||
| XM_005267834.1 | XP_005267891.1 | 6 | 2125 | 76 | ||
| XM_017021426.1 | XP_016876915.1 | 8 | 2464 | 76 | ||
| GenBank mRNA | AK002136.1 | - | 7 | 2299 | 76 | |
| AK311104.1 | - | 2 | 987 | 76 | ||
| AY358687.1 | - | 7 | 1769 | 76 | ||
More Information
Set Membership: |
|

SLC39A9
Genomic Map