Assay Details
| Gene Symbol: IGHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF067420.1 | - | 4 | 712 | 63 | |
| AF283666.1 | - | 5 | 684 | 63 | ||
| AJ294729.1 | - | 1 | 224 | 63 | ||
| AK027379.1 | - | 3 | 723 | 63 | ||
| AK058027.1 | - | 4 | 714 | 63 | ||
| AK074651.1 | - | 4 | 723 | 63 | ||
| AK092384.1 | - | 5 | 711 | 63 | ||
| AK092803.1 | - | 3 | 768 | 63 | ||
| AK092819.1 | - | 4 | 927 | 63 | ||
| AK093721.1 | - | 3 | 735 | 63 | ||
| AK095222.1 | - | 3 | 706 | 63 | ||
| AK096985.1 | - | 3 | 711 | 63 | ||
| AK097017.1 | - | 4 | 726 | 63 | ||
| AK097028.1 | - | 4 | 716 | 63 | ||
| AK097076.1 | - | 5 | 729 | 63 | ||
| AK097303.1 | - | 4 | 736 | 63 | ||
| AK097305.1 | - | 4 | 729 | 63 | ||
| AK097320.1 | - | 5 | 745 | 63 | ||
| AK097329.1 | - | 4 | 718 | 63 | ||
| AK097345.1 | - | 4 | 726 | 63 | ||
| AK097348.1 | - | 4 | 697 | 63 | ||
| AK097362.1 | - | 4 | 677 | 63 | ||
| AK097955.1 | - | 4 | 743 | 63 | ||
| AK123546.1 | - | 5 | 727 | 63 | ||
| AK123783.1 | - | 3 | 742 | 63 | ||
| AK125238.1 | - | 3 | 711 | 63 | ||
| AK125582.1 | - | 4 | 736 | 63 | ||
| AK126279.1 | - | 5 | 735 | 63 | ||
| AK126543.1 | - | 4 | 700 | 63 | ||
| AK127921.1 | - | 4 | 682 | 63 | ||
| AK128476.1 | - | 5 | 720 | 63 | ||
| AK128565.1 | - | 4 | 721 | 63 | ||
| AK128652.1 | - | 5 | 2704 | 63 | ||
| AK128664.1 | - | 4 | 741 | 63 | ||
| AK129620.1 | - | 4 | 689 | 63 | ||
| AK130433.1 | - | 6 | 729 | 63 | ||
| AK130453.1 | - | 4 | 750 | 63 | ||
| AK130465.1 | - | 4 | 702 | 63 | ||
| AK130468.1 | - | 4 | 759 | 63 | ||
| AK130476.1 | - | 4 | 742 | 63 | ||
| AK130501.1 | - | 4 | 729 | 63 | ||
| AK130529.1 | - | 4 | 703 | 63 | ||
| AK130796.1 | - | 4 | 693 | 63 | ||
| AK131065.1 | - | 4 | 723 | 63 | ||
| AY885214.2 | - | 1 | 308 | 63 | ||
| AY885215.2 | - | 1 | 308 | 63 | ||
| BC005951.1 | - | 4 | 719 | 63 | ||
| BC016369.1 | - | 4 | 695 | 63 | ||
| BC032249.1 | - | 5 | 724 | 63 | ||
| BC073771.1 | - | 5 | 700 | 63 | ||
| BC087841.1 | - | 4 | 671 | 63 | ||
| BC092449.1 | - | 4 | 664 | 63 | ||
| BX640624.1 | - | 5 | 723 | 63 | ||
| BX640626.1 | - | 5 | 683 | 63 | ||
| BX640847.1 | - | 3 | 684 | 63 | ||
| Gene Symbol: IGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123975.1 | - | 4 | 63 | ||
| AK125079.1 | - | 3 | 63 | |||
| AK126352.1 | - | 4 | 63 | |||
| BC066594.1 | - | 4 | 63 | |||
| BX538066.1 | - | 5 | 63 | |||
| BX640822.1 | - | 4 | 63 | |||
| BX640854.1 | - | 4 | 63 | |||
| Gene Symbol: IGHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL389978.1 | - | 3 | 63 | ||
| BC073765.1 | - | 3 | 63 | |||
| BX538077.1 | - | 4 | 63 | |||
| BX640625.1 | - | 4 | 63 | |||
| BX640710.1 | - | 3 | 63 | |||
More Information
Set Membership: |
|

IGHA1
Genomic Map