Assay Details
| Gene Symbol: IGHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF067420.1 | - | 6 | 1347 | 131 | |
| AF283666.1 | - | 7 | 1319 | 131 | ||
| AJ294729.1 | - | 3 | 859 | 131 | ||
| AK027379.1 | - | 5 | 1358 | 131 | ||
| AK058027.1 | - | 6 | 1349 | 131 | ||
| AK074651.1 | - | 6 | 1358 | 131 | ||
| AK092384.1 | - | 7 | 1346 | 131 | ||
| AK092803.1 | - | 5 | 1403 | 131 | ||
| AK092819.1 | - | 6 | 1562 | 131 | ||
| AK093721.1 | - | 5 | 1370 | 131 | ||
| AK095222.1 | - | 5 | 1341 | 131 | ||
| AK096985.1 | - | 5 | 1346 | 131 | ||
| AK097017.1 | - | 6 | 1361 | 131 | ||
| AK097028.1 | - | 6 | 1351 | 131 | ||
| AK097076.1 | - | 7 | 1364 | 131 | ||
| AK097305.1 | - | 6 | 1364 | 131 | ||
| AK097320.1 | - | 7 | 1380 | 131 | ||
| AK097329.1 | - | 6 | 1353 | 131 | ||
| AK097345.1 | - | 6 | 1361 | 131 | ||
| AK097348.1 | - | 6 | 1332 | 131 | ||
| AK097362.1 | - | 6 | 1312 | 131 | ||
| AK097955.1 | - | 6 | 1378 | 131 | ||
| AK123546.1 | - | 7 | 1362 | 131 | ||
| AK123783.1 | - | 5 | 1377 | 131 | ||
| AK125238.1 | - | 5 | 1346 | 131 | ||
| AK125582.1 | - | 6 | 1371 | 131 | ||
| AK126279.1 | - | 7 | 1370 | 131 | ||
| AK126543.1 | - | 6 | 1335 | 131 | ||
| AK127921.1 | - | 6 | 1317 | 131 | ||
| AK128476.1 | - | 7 | 1355 | 131 | ||
| AK128565.1 | - | 6 | 1356 | 131 | ||
| AK128652.1 | - | 7 | 3339 | 131 | ||
| AK128664.1 | - | 6 | 1376 | 131 | ||
| AK129592.1 | - | 6 | 1352 | 131 | ||
| AK129620.1 | - | 6 | 1324 | 131 | ||
| AK130433.1 | - | 8 | 1364 | 131 | ||
| AK130453.1 | - | 6 | 1385 | 131 | ||
| AK130465.1 | - | 6 | 1337 | 131 | ||
| AK130468.1 | - | 6 | 1394 | 131 | ||
| AK130476.1 | - | 6 | 1377 | 131 | ||
| AK130501.1 | - | 6 | 1364 | 131 | ||
| AK130529.1 | - | 6 | 1338 | 131 | ||
| AK130796.1 | - | 6 | 1328 | 131 | ||
| AK131065.1 | - | 6 | 1356 | 131 | ||
| BC005951.1 | - | 6 | 1354 | 131 | ||
| BC016369.1 | - | 6 | 1330 | 131 | ||
| BC032249.1 | - | 7 | 1359 | 131 | ||
| BC073771.1 | - | 7 | 1335 | 131 | ||
| BC087841.1 | - | 6 | 1306 | 131 | ||
| BC092449.1 | - | 6 | 1299 | 131 | ||
| BX640624.1 | - | 7 | 1358 | 131 | ||
| BX640626.1 | - | 7 | 1318 | 131 | ||
| BX640847.1 | - | 5 | 1319 | 131 | ||
| Gene Symbol: IGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123975.1 | - | 6 | 1340 | 131 | |
| AK125079.1 | - | 5 | 1303 | 131 | ||
| AK126352.1 | - | 6 | 1286 | 131 | ||
| BC066594.1 | - | 6 | 1249 | 131 | ||
| BX640822.1 | - | 6 | 1329 | 131 | ||
| BX640854.1 | - | 6 | 1317 | 131 | ||
| Gene Symbol: IGHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL389978.1 | - | 5 | 1050 | 131 | |
| BC073765.1 | - | 5 | 1257 | 131 | ||
| BX538077.1 | - | 6 | 1278 | 131 | ||
| BX640625.1 | - | 6 | 1300 | 131 | ||
| BX640710.1 | - | 5 | 1295 | 131 | ||
More Information
Set Membership: |
|

IGHA1
Genomic Map