Assay Details
| Gene Symbol: LOC101928042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135833.1 | 1 | 255 | 100 | ||
| Gene Symbol: LOC101927579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522275.1 | XP_011520577.1 | 1 | 652 | 100 | |
| XM_011522282.2 | XP_011520584.2 | 1 | 652 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011522275.1, XM_011522282.2 transcripts.
Set Membership: |
|

LOC101928042
Genomic Map