Assay Details
| Gene Symbol: PRR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018304.3 | NP_060774.2 | 10 | 2545 | 97 | |
| GenBank mRNA | AK000296.1 | - | 7 | 1861 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR11| Gene Symbol: PRR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018304.3 | NP_060774.2 | 10 | 2545 | 97 | |
| GenBank mRNA | AK000296.1 | - | 7 | 1861 | 97 | |
Set Membership: |
|