Assay Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011524620.2 | XP_011522922.1 | 7 | 2093 | 78 | |
| XM_011524624.2 | XP_011522926.1 | 5 | 1931 | 78 | ||
| XM_017024459.1 | XP_016879948.1 | 6 | 2006 | 78 | ||
| XM_017024460.1 | XP_016879949.1 | 7 | 2104 | 78 | ||
More Information
Set Membership: |
|

TMEM235
Genomic Map