Assay Details
| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270614.1 | NP_001257543.1 | 5 | 1720 | 80 | |
| NM_001270615.1 | NP_001257544.1 | 5 | 1787 | 80 | ||
| NM_133328.3 | NP_579874.1 | 5 | 1802 | 80 | ||
| NR_073046.1 | 5 | 1534 | 80 | |||
| NR_073047.1 | 4 | 1354 | 80 | |||
| NR_073048.1 | 4 | 1436 | 80 | |||
| NR_073049.1 | 5 | 1458 | 80 | |||
| NR_073050.1 | 4 | 1421 | 80 | |||
| NR_073051.1 | 5 | 1520 | 80 | |||
| NR_073052.1 | 3 | 1295 | 80 | |||
| XM_011526569.1 | XP_011524871.1 | 7 | 1846 | 80 | ||
| XM_011526571.1 | XP_011524873.1 | 6 | 1824 | 80 | ||
| XM_011526572.1 | XP_011524874.1 | 6 | 1748 | 80 | ||
| XM_017026402.1 | XP_016881891.1 | 7 | 1840 | 80 | ||
| XM_017026403.1 | XP_016881892.1 | 7 | 1908 | 80 | ||
| GenBank mRNA | AA635978.1 | - | 1 | 80 | ||
| AK075328.1 | - | 6 | 1667 | 80 | ||
| AK130203.1 | - | 5 | 1442 | 80 | ||
| AY125488.1 | - | 5 | 1671 | 80 | ||
| BC013372.2 | - | 5 | 1664 | 3941907 | 80 | |
| BC027930.1 | - | 5 | 1671 | 5199772 | 80 | |
More Information
Set Membership: |
|

DEDD2
Genomic Map