Assay Details
| Gene Symbol: EXOC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321729.1 | NP_001308658.1 | 23 | 3754 | 86 | |
| NM_001321730.1 | NP_001308659.1 | 22 | 3619 | 86 | ||
| NM_001321731.1 | NP_001308660.1 | 23 | 3795 | 86 | ||
| NM_001321733.1 | NP_001308662.1 | 21 | 3607 | 86 | ||
| NM_001321734.1 | NP_001308663.1 | 22 | 3588 | 86 | ||
| NM_015189.2 | NP_056004.1 | 22 | 3742 | 86 | ||
| NR_135773.1 | 22 | 3680 | 86 | |||
| NR_135774.1 | 20 | 3420 | 86 | |||
| XM_005264224.1 | XP_005264281.1 | 16 | 3046 | 86 | ||
| XM_017003641.1 | XP_016859130.1 | 19 | 3188 | 86 | ||
| XM_017003642.1 | XP_016859131.1 | 17 | 2801 | 86 | ||
| GenBank mRNA | AK023791.1 | - | 10 | 2303 | 86 | |
More Information
Set Membership: |
|

EXOC6B
Genomic Map