Assay Details
| Gene Symbol: FAHD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011510745.2 | XP_011509047.1 | 10 | 96 | ||
| XM_011510746.2 | XP_011509048.1 | 10 | 96 | |||
| XM_011510747.2 | XP_011509049.1 | 9 | 96 | |||
| XM_011510748.2 | XP_011509050.1 | 10 | 96 | |||
| XM_017003470.1 | XP_016858959.1 | 10 | 96 | |||
| XM_017003471.1 | XP_016858960.1 | 10 | 96 | |||
| XM_017003472.1 | XP_016858961.1 | 9 | 96 | |||
| Gene Symbol: LOC653924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC104425.1 | - | 1 | 96 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC104424.1 | - | 505 | Chr. - - [-] on Build | 96 | ||
More Information
Set Membership: |
|

FAHD2B
Genomic Map