Assay Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015348.1 | NP_056163.1 | 41 | 6319 | 114 | |
| XM_005263909.1 | XP_005263966.1 | 42 | 6405 | 114 | ||
| XM_005263910.1 | XP_005263967.1 | 42 | 6402 | 114 | ||
| XM_005263911.1 | XP_005263968.1 | 41 | 6288 | 114 | ||
| XM_005263912.3 | XP_005263969.1 | 40 | 6493 | 114 | ||
| XM_011510869.1 | XP_011509171.1 | 41 | 6219 | 114 | ||
| GenBank mRNA | AK025852.1 | - | 11 | 1967 | 114 | |
| AK098792.1 | - | 10 | 2142 | 114 | ||
| BC018018.1 | - | 2 | 624 | 114 | ||
| BC036841.1 | - | 5 | 1484 | 114 | ||
| BX647728.1 | - | 2 | 7148 | 114 | ||
| D87446.1 | - | 40 | 5857 | 114 | ||
More Information
Set Membership: |
|

TMEM131
Genomic Map