Assay Details
| Gene Symbol: GACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126370.1 | 2 - 3 | 283 | 124 | ||
| GenBank mRNA | DB096621.1 | - | 2 - 3 | 283 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GACAT1| Gene Symbol: GACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126370.1 | 2 - 3 | 283 | 124 | ||
| GenBank mRNA | DB096621.1 | - | 2 - 3 | 283 | 124 | |
Set Membership: |
|