Assay Details
Gene Symbol: ESR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122742.1 | NP_001116214.1 | 2 | 216 | 71 | |
XM_006715374.3 | XP_006715437.1 | 2 | 216 | 71 | ||
XM_011535543.2 | XP_011533845.1 | 2 | 1927 | 71 | ||
XM_011535547.2 | XP_011533849.1 | 2 | 216 | 71 | ||
XM_017010376.1 | XP_016865865.1 | 2 | 2663 | 71 | ||
XM_017010377.1 | XP_016865866.1 | 3 | 2722 | 71 | ||
XM_017010378.1 | XP_016865867.1 | 2 | 174 | 71 | ||
XM_017010379.1 | XP_016865868.1 | 2 | 208 | 71 | ||
GenBank mRNA | AB588629.1 | - | 2 | 90 | 71 | |
AB588630.1 | - | 2 | 90 | 71 | ||
AB685815.1 | - | 2 | 116 | 71 | ||
AB685816.1 | - | 3 | 164 | 71 | ||
AB685817.1 | - | 3 | 175 | 71 | ||
AB685819.1 | - | 2 | 116 | 71 | ||
AB685820.1 | - | 3 | 175 | 71 | ||
AK309580.1 | - | 2 | 114 | 71 | ||
DA329077.1 | - | 2 | 216 | 71 | ||
U68068.1 | - | 2 | 132 | 71 | ||
X86816.1 | - | 2 | 120 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB588629.1, AB588630.1, AB685815.1, AB685816.1, AB685817.1, AB685819.1, AB685820.1, NM_001122742.1, U68068.1, XM_006715374.3, XM_011535543.2, XM_011535547.2, XM_017010376.1, XM_017010377.1, XM_017010378.1, XM_017010379.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
