Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184892.1 | NP_001171821.1 | 3 | 788 | 89 | |
| GenBank mRNA | AF218022.1 | - | 3 | 715 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF218022.1, NM_001184892.1 transcripts.
Set Membership: |
|

FLAD1
Genomic Map