Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530007.2 | XP_011528309.1 | 1 | 841 | 71 | |
| XM_011530009.2 | XP_011528311.1 | 1 | 841 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530007.2, XM_011530009.2 transcripts.
Set Membership: |
|

HIC2
Genomic Map