Assay Details
| Gene Symbol: FAM120B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286379.1 | NP_001273308.1 | 2 | 1900 | 115 | |
| NM_001286380.1 | NP_001273309.1 | 2 | 1663 | 115 | ||
| NM_032448.2 | NP_115824.1 | 2 | 1651 | 115 | ||
| XM_011536185.2 | XP_011534487.1 | 2 | 1645 | 115 | ||
| XM_017011358.1 | XP_016866847.1 | 2 | 1645 | 115 | ||
| XM_017011359.1 | XP_016866848.1 | 2 | 1645 | 115 | ||
| XM_017011360.1 | XP_016866849.1 | 2 | 1645 | 115 | ||
| GenBank mRNA | AB058741.1 | - | 2 | 1564 | 115 | |
| AK056435.1 | - | 2 | 1630 | 115 | ||
| AK294415.1 | - | 2 | 1900 | 115 | ||
| AK296730.1 | - | 3 | 1270 | 115 | ||
| AK299890.1 | - | 2 | 1663 | 115 | ||
| AL832883.1 | - | 2 | 1549 | 115 | ||
| BC012177.1 | - | 2 | 1617 | 4650497 | 115 | |
| DQ873695.1 | - | 2 | 1652 | 115 | ||
More Information
Set Membership: |
|

FAM120B
Genomic Map