Assay Details
| Gene Symbol: RTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321859.1 | NP_001308788.1 | 9 | 3620 | 141 | |
| NM_001321860.1 | NP_001308789.1 | 9 | 3690 | 141 | ||
| NM_001321861.1 | NP_001308790.1 | 9 | 3653 | 141 | ||
| NM_001321862.1 | NP_001308791.1 | 9 | 3640 | 141 | ||
| NM_001321863.1 | NP_001308792.1 | 9 | 3590 | 141 | ||
| NM_001321904.1 | NP_001308833.1 | 10 | 4068 | 141 | ||
| NM_007008.2 | NP_008939.1 | 7 | 1310 | 141 | ||
| NM_020532.4 | NP_065393.1 | 9 | 4373 | 141 | ||
| NM_153828.2 | NP_722550.1 | 7 | 1916 | 141 | ||
| NM_207520.1 | NP_997403.1 | 8 | 1973 | 141 | ||
| NM_207521.1 | NP_997404.1 | 9 | 3631 | 141 | ||
| NR_135829.1 | 7 | 1196 | 141 | |||
| NR_135830.1 | 7 | 1174 | 141 | |||
| XM_005264434.3 | XP_005264491.1 | 10 | 3767 | 141 | ||
| XM_017004518.1 | XP_016860007.1 | 10 | 3806 | 141 | ||
| XM_017004519.1 | XP_016860008.1 | 10 | 3923 | 141 | ||
| GenBank mRNA | AF077050.1 | - | 7 | 1306 | 141 | |
| AF087901.1 | - | 8 | 1137 | 141 | ||
| AF125103.1 | - | 7 | 1310 | 141 | ||
| AF132047.1 | - | 7 | 1793 | 141 | ||
| AF132048.1 | - | 7 | 1208 | 141 | ||
| AF148537.1 | - | 10 | 4152 | 141 | ||
| AF148538.1 | - | 7 | 1755 | 141 | ||
| AF177332.1 | - | 7 | 1211 | 141 | ||
| AK172726.1 | - | 5 | 736 | 141 | ||
| AY102276.1 | - | 7 | 1326 | 141 | ||
| AY102277.1 | - | 7 | 1858 | 141 | ||
| AY102278.1 | - | 8 | 1915 | 141 | ||
| AY102279.1 | - | 9 | 4315 | 141 | ||
| AY123245.1 | - | 9 | 3628 | 141 | ||
| AY123246.1 | - | 9 | 3686 | 141 | ||
| AY123247.1 | - | 9 | 3649 | 141 | ||
| AY123248.1 | - | 9 | 3635 | 141 | ||
| AY123249.1 | - | 9 | 3596 | 141 | ||
| AY123250.1 | - | 9 | 3586 | 141 | ||
| BC014366.1 | - | 7 | 1204 | 141 | ||
| BC150182.1 | - | 9 | 4324 | 40148210 | 141 | |
| BC152555.1 | - | 9 | 4324 | 40148984 | 141 | |
| Gene Symbol: EML6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017004098.1 | XP_016859587.1 | 43 | 141 | ||
| XM_017004099.1 | XP_016859588.1 | 42 | 141 | |||
| XM_017004101.1 | XP_016859590.1 | 37 | 141 | |||
More Information
Set Membership: |
|

RTN4
Genomic Map