Assay Details
| Gene Symbol: CR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006658.2 | NP_001006659.1 | 11 | 2227 | 73 | |
| XM_011509206.2 | XP_011507508.1 | 11 | 2123 | 73 | ||
| GenBank mRNA | AK223627.1 | - | 11 | 2199 | 73 | |
| AK301496.1 | - | 11 | 2195 | 73 | ||
| BC136394.1 | - | 11 | 2201 | 9020381 | 73 | |
| J03565.1 | - | 11 | 2171 | 73 | ||
| M26317.1 | - | 2 | 156 | 73 | ||
More Information
Set Membership: |
|

CR2
Genomic Map