Assay Details
| Gene Symbol: DLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001931.4 | NP_001922.2 | 1 | 633 | 55 | |
| XM_011542647.2 | XP_011540949.1 | 1 | 280 | 55 | ||
| GenBank mRNA | AF317200.1 | - | 1 | 22 | 55 | |
| AK057299.1 | - | 1 | 633 | 55 | ||
| AK223596.1 | - | 1 | 22 | 55 | ||
| AK296274.1 | - | 1 | 39 | 55 | ||
| AK297103.1 | - | 1 | 36 | 55 | ||
| AK303930.1 | - | 1 | 39 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF317200.1, AK057299.1, AK296274.1, AK297103.1, AK303930.1, NM_001931.4, XM_011542647.2 transcripts.
Set Membership: |
|

DLAT
Genomic Map