Assay Details
| Gene Symbol: RELA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145138.1 | NP_001138610.1 | 9 | 1052 | 70 | |
| NM_001243984.1 | NP_001230913.1 | 9 | 1061 | 70 | ||
| NM_001243985.1 | NP_001230914.1 | 9 | 1061 | 70 | ||
| NM_021975.3 | NP_068810.3 | 9 | 1061 | 70 | ||
| XM_011545206.1 | XP_011543508.1 | 8 | 829 | 70 | ||
| XM_011545207.1 | XP_011543509.1 | 7 | 724 | 70 | ||
| GenBank mRNA | AK303262.1 | - | 9 | 1040 | 70 | |
| AK308020.1 | - | 9 | 1005 | 70 | ||
| AK308478.1 | - | 8 | 1107 | 70 | ||
| BC011603.1 | - | 9 | 802 | 70 | ||
| BC033522.1 | - | 8 | 918 | 5162908 | 70 | |
| BC110830.1 | - | 9 | 984 | 6019711 | 70 | |
| L19067.1 | - | 8 | 920 | 70 | ||
| M62399.1 | - | 9 | 1000 | 70 | ||
| U33838.1 | - | 2 | 122 | 70 | ||
More Information
Set Membership: |
|

RELA
Genomic Map