Assay Details
| Gene Symbol: HLA-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002127.5 | NP_002118.1 | 4 | 711 | 68 | |
| XM_017010816.1 | XP_016866305.1 | 4 | 620 | 68 | ||
| GenBank mRNA | AF071017.1 | - | 2 | 327 | 68 | |
| AF071018.1 | - | 2 | 327 | 68 | ||
| AF071019.1 | - | 2 | 327 | 68 | ||
| AF071020.1 | - | 2 | 327 | 68 | ||
| AF196177.1 | - | 3 | 532 | 68 | ||
| AF196178.1 | - | 3 | 532 | 68 | ||
| AF226990.2 | - | 3 | 557 | 68 | ||
| AF436089.1 | - | 2 | 460 | 68 | ||
| AK093478.1 | - | 3 | 827 | 68 | ||
| AK302054.1 | - | 4 | 637 | 68 | ||
| AY359818.1 | - | 3 | 535 | 68 | ||
| BC021708.2 | - | 4 | 620 | 4822521 | 68 | |
| M32800.1 | - | 3 | 533 | 68 | ||
| M90683.1 | - | 3 | 533 | 68 | ||
| U58024.1 | - | 2 | 460 | 68 | ||
| U58025.1 | - | 2 | 460 | 68 | ||
| U58026.1 | - | 2 | 460 | 68 | ||
| U58027.1 | - | 2 | 460 | 68 | ||
| U58028.1 | - | 2 | 460 | 68 | ||
| U58029.1 | - | 2 | 460 | 68 | ||
| U58094.1 | - | 2 | 460 | 68 | ||
| U88244.1 | - | 2 | 432 | 68 | ||
| X17273.1 | - | 3 | 538 | 68 | ||
More Information
Set Membership: |
|

HLA-G
Genomic Map