Assay Details
| Gene Symbol: MYO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300899.1 | NP_001287828.1 | 8 | 884 | 74 | |
| NM_004999.3 | NP_004990.3 | 8 | 868 | 74 | ||
| XM_005248719.3 | XP_005248776.1 | 8 | 884 | 74 | ||
| XM_005248720.3 | XP_005248777.1 | 8 | 884 | 74 | ||
| XM_005248721.3 | XP_005248778.1 | 8 | 884 | 74 | ||
| XM_005248722.3 | XP_005248779.1 | 8 | 884 | 74 | ||
| XM_005248724.3 | XP_005248781.1 | 8 | 884 | 74 | ||
| XM_005248726.3 | XP_005248783.1 | 8 | 884 | 74 | ||
| XM_017010899.1 | XP_016866388.1 | 8 | 884 | 74 | ||
| GenBank mRNA | AB002387.1 | - | 8 | 728 | 74 | |
| BC146764.1 | - | 8 | 728 | 9007206 | 74 | |
| BX647397.1 | - | 8 | 859 | 74 | ||
| L29145.1 | - | 3 | 154 | 74 | ||
| U90236.2 | - | 7 | 709 | 74 | ||
More Information
Set Membership: |
|

MYO6
Genomic Map