Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000602.4 | NP_000593.1 | 2 | 241 | 109 | |
| XM_017012260.1 | XP_016867749.1 | 2 | 243 | 109 | ||
| GenBank mRNA | BC010860.1 | - | 2 | 206 | 3893914 | 109 |
| CN309001.1 | - | 2 | 242 | 109 | ||
| M16006.1 | - | 2 | 159 | 109 | ||
| X04429.1 | - | 2 | 210 | 109 | ||
| X04729.1 | - | 2 | 227 | 109 | ||
| X12701.1 | - | 2 | 210 | 109 | ||
More Information
Set Membership: |
|

SERPINE1
Genomic Map