Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000739.2 | NP_000730.1 | 2 | 161 | 77 | |
| NM_001006626.1 | NP_001006627.1 | 2 | 124 | 77 | ||
| NM_001006627.1 | NP_001006628.1 | 2 | 413 | 77 | ||
| NM_001006628.1 | NP_001006629.1 | 1 | 87 | 77 | ||
| NM_001006629.1 | NP_001006630.1 | 1 | 96 | 77 | ||
| NM_001006630.1 | NP_001006631.1 | 2 | 413 | 77 | ||
| NM_001006631.1 | NP_001006632.1 | 2 | 105 | 77 | ||
| NM_001006632.1 | NP_001006633.1 | 2 | 161 | 77 | ||
| GenBank mRNA | AK125233.1 | - | 2 | 161 | 77 | |
| AK290236.1 | - | 2 | 77 | |||
| AL832585.1 | - | 2 | 397 | 77 | ||
| AY034603.2 | - | 2 | 411 | 77 | ||
| AY034604.2 | - | 2 | 161 | 77 | ||
| AY034605.2 | - | 1 | 87 | 77 | ||
| AY219703.1 | - | 1 | 96 | 77 | ||
| AY219704.1 | - | 2 | 413 | 77 | ||
| AY219705.1 | - | 2 | 124 | 77 | ||
| AY456125.1 | - | 2 | 161 | 77 | ||
| AY456126.1 | - | 2 | 105 | 77 | ||
| AY456127.1 | - | 2 | 105 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK290236.1, AY034603.2, AY034604.2, AY034605.2, AY219703.1, AY219704.1, AY219705.1, AY456125.1, AY456126.1, AY456127.1, NM_000739.2, NM_001006626.1, NM_001006627.1, NM_001006628.1, NM_001006629.1, NM_001006630.1, NM_001006631.1, NM_001006632.1 transcripts.
Set Membership: |
|

CHRM2
Genomic Map