Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000739.2 | NP_000730.1 | 1 | 29 | 53 | |
| NM_001006632.1 | NP_001006633.1 | 1 | 29 | 53 | ||
| GenBank mRNA | AK125233.1 | - | 1 | 29 | 53 | |
| AY034604.2 | - | 1 | 29 | 53 | ||
| AY456125.1 | - | 1 | 29 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY034604.2, AY456125.1, NM_000739.2, NM_001006632.1 transcripts.
Set Membership: |
|

CHRM2
Genomic Map