Assay Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271004.1 | NP_001257933.1 | 2 | 435 | 82 | |
| NM_006528.3 | NP_006519.1 | 2 | 435 | 82 | ||
| GenBank mRNA | AK024817.1 | - | 2 | 197 | 82 | |
| AK075425.1 | - | 2 | 197 | 82 | ||
| AK075427.1 | - | 2 | 196 | 82 | ||
| AK129833.1 | - | 2 | 197 | 82 | ||
| AK313260.1 | - | 2 | 197 | 82 | ||
| AY691946.1 | - | 2 | 173 | 82 | ||
| BC005330.1 | - | 2 | 197 | 3929933 | 82 | |
| D29992.1 | - | 2 | 178 | 82 | ||
| DA831865.1 | - | 2 | 435 | 82 | ||
| DA833200.1 | - | 2 | 196 | 82 | ||
| EF488667.1 | - | 1 | 36 | 82 | ||
| L27624.1 | - | 2 | 160 | 82 | ||
More Information
Set Membership: |
|

TFPI2
Genomic Map