Assay Details
| Gene Symbol: SET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122821.1 | NP_001116293.1 | 1 | 222 | 98 | |
| XM_017015013.1 | XP_016870502.1 | 1 | 438 | 98 | ||
| XM_017015014.1 | XP_016870503.1 | 2 | 117 | 98 | ||
| XM_017015015.1 | XP_016870504.1 | 1 | 112 | 98 | ||
| GenBank mRNA | AK300794.1 | - | 1 | 81 | 98 | |
| CB129868.1 | - | 1 | 222 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300794.1, NM_001122821.1, XM_017015013.1, XM_017015014.1, XM_017015015.1 transcripts.
Set Membership: |
|

SET
Genomic Map