Assay Details
| Gene Symbol: PLS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136025.4 | NP_001129497.1 | 4 | 441 | 67 | |
| NM_001172335.2 | NP_001165806.1 | 4 | 360 | 67 | ||
| NM_001282337.1 | NP_001269266.1 | 5 | 845 | 67 | ||
| NM_001282338.1 | NP_001269267.1 | 5 | 823 | 67 | ||
| NM_005032.6 | NP_005023.2 | 4 | 726 | 67 | ||
| XM_011537534.1 | XP_011535836.1 | 4 | 417 | 67 | ||
| GenBank mRNA | AK222801.1 | - | 4 | 382 | 67 | |
| AK294509.1 | - | 5 | 467 | 67 | ||
| AK298501.1 | - | 4 | 402 | 67 | ||
| AK298530.1 | - | 4 | 348 | 67 | ||
| AK300575.1 | - | 5 | 777 | 67 | ||
| AK311397.1 | - | 5 | 786 | 67 | ||
| AK312391.1 | - | 4 | 400 | 67 | ||
| BC039049.1 | - | 4 | 340 | 3446726 | 67 | |
| BC056898.1 | - | 4 | 444 | 6064540 | 67 | |
| BP214207.1 | - | 4 | 440 | 67 | ||
| DC357886.1 | - | 4 | 441 | 67 | ||
| M22299.1 | - | 2 | 127 | 67 | ||
| M34427.1 | - | 4 | 394 | 67 | ||
More Information
Set Membership: |
|

PLS3
Genomic Map