Assay Details
| Gene Symbol: LRRC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135217.1 | NP_001128689.1 | 4 | 774 | 76 | |
| NM_006992.3 | NP_008923.1 | 4 | 602 | 76 | ||
| NM_201650.2 | NP_964013.1 | 4 | 602 | 76 | ||
| GenBank mRNA | AK223535.1 | - | 4 | 536 | 76 | |
| AK292291.1 | - | 4 | 537 | 76 | ||
| AK292367.1 | - | 4 | 602 | 76 | ||
| AY834277.1 | - | 3 | 382 | 76 | ||
| BC029858.1 | - | 4 | 5170643 | 76 | ||
| DB027456.1 | - | 3 | 534 | 76 | ||
| U72508.1 | - | 4 | 494 | 76 | ||
| U72509.1 | - | 2 | 209 | 76 | ||
| U72510.1 | - | 3 | 76 | |||
More Information
Set Membership: |
|

LRRC23
Genomic Map