Assay Details
| Gene Symbol: DDX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020414.3 | NP_065147.1 | 2 | 239 | 95 | |
| GenBank mRNA | AB209382.1 | - | 1 | 159 | 95 | |
| AF161446.1 | - | 2 | 194 | 95 | ||
| AF214731.1 | - | 2 | 238 | 95 | ||
| AK025162.1 | - | 2 | 235 | 95 | ||
| AK294613.1 | - | 2 | 193 | 95 | ||
| AK296726.1 | - | 2 | 229 | 95 | ||
| AL136886.1 | - | 2 | 215 | 95 | ||
| BC008847.2 | - | 2 | 215 | 4301025 | 95 | |
| BC009406.2 | - | 2 | 215 | 4303448 | 95 | |
| BC096826.1 | - | 2 | 210 | 30925011 | 95 | |
| BX537533.1 | - | 2 | 226 | 95 | ||
More Information
Set Membership: |
|

DDX24
Genomic Map