Assay Details
| Gene Symbol: CCL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006273.3 | NP_006264.2 | 2 | 205 | 80 | |
| GenBank mRNA | AF043338.1 | - | 1 | 61 | 80 | |
| BC092436.1 | - | 2 | 199 | 30418073 | 80 | |
| BC112258.1 | - | 2 | 195 | 8327726 | 80 | |
| BC112260.1 | - | 2 | 195 | 8327728 | 80 | |
| CD523198.1 | - | 2 | 208 | 80 | ||
| X71087.1 | - | 2 | 200 | 80 | ||
More Information
Set Membership: |
|

CCL7
Genomic Map