Assay Details
| Gene Symbol: SAMD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054643.1 | - | 7 | 1314 | 92 | |
| JX093066.1 | - | 12 | 1787 | 92 | ||
| JX093080.1 | - | 11 | 1981 | 92 | ||
| JX093083.1 | - | 11 | 1978 | 92 | ||
| JX093091.1 | - | 5 | 628 | 92 | ||
More Information
Set Membership: |
|

SAMD11
Genomic Map