Assay Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306090.1 | NP_001293019.1 | 21 | 3754 | 108 | |
| NM_153717.2 | NP_714928.1 | 21 | 3757 | 108 | ||
| GenBank mRNA | AF216184.1 | - | 21 | 3757 | 108 | |
| AF216185.1 | - | 21 | 3757 | 108 | ||
More Information
Set Membership: |
|

EVC
Genomic Map