Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000947.4 | NP_000938.2 | 2 - 3 | 249 | 108 | |
| NM_001282487.1 | NP_001269416.1 | 2 - 3 | 249 | 108 | ||
| NM_001282488.1 | NP_001269417.1 | 2 - 3 | 280 | 108 | ||
| XM_017011004.1 | XP_016866493.1 | 2 - 3 | 249 | 108 | ||
| XM_017011005.1 | XP_016866494.1 | 2 - 3 | 249 | 108 | ||
| XM_017011006.1 | XP_016866495.1 | 2 - 3 | 249 | 108 | ||
| XM_017011007.1 | XP_016866496.1 | 1 - 2 | 164 | 108 | ||
| GenBank mRNA | AK223287.1 | - | 2 - 3 | 238 | 108 | |
| AK291915.1 | - | 2 - 3 | 236 | 108 | ||
| BC017833.1 | - | 2 - 3 | 249 | 108 | ||
| BC064931.1 | - | 2 - 3 | 234 | 6148494 | 108 | |
| BI547222.1 | - | 2 - 3 | 285 | 108 | ||
| X74331.1 | - | 2 - 3 | 242 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011007.1 transcripts.
Set Membership: |
|

PRIM2
Genomic Map