Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002901.3 | NP_001002901.1 | 5 - 6 | 546 | 76 | |
| NM_001288829.1 | NP_001275758.1 | 3 - 4 | 332 | 76 | ||
| NM_001288830.1 | NP_001275759.1 | 2 - 3 | 311 | 76 | ||
| NM_001288831.1 | NP_001275760.1 | 3 - 4 | 332 | 76 | ||
| NM_001288832.1 | NP_001275761.1 | 2 - 3 | 311 | 76 | ||
| NM_001320241.1 | NP_001307170.1 | 4 - 5 | 360 | 76 | ||
| GenBank mRNA | AK290733.1 | - | 4 - 5 | 360 | 76 | |
| AY670683.1 | - | 3 - 4 | 327 | 76 | ||
| AY670684.1 | - | 3 - 4 | 327 | 76 | ||
| AY670685.1 | - | 2 - 3 | 306 | 76 | ||
| AY670686.1 | - | 3 - 4 | 327 | 76 | ||
| AY670687.1 | - | 2 - 3 | 306 | 76 | ||
| AY786314.1 | - | 3 - 4 | 312 | 76 | ||
| BX364259.2 | - | 4 - 5 | 502 | 76 | ||
| DQ021957.1 | - | 5 - 6 | 546 | 76 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map