Assay Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290030.1 | NP_001276959.1 | 2 - 3 | 541 | 80 | |
| NM_001290031.1 | NP_001276960.1 | 2 - 3 | 541 | 80 | ||
| XM_011510998.2 | XP_011509300.1 | 4 - 5 | 673 | 80 | ||
| XM_011510999.2 | XP_011509301.1 | 3 - 4 | 340 | 80 | ||
| XM_017003875.1 | XP_016859364.1 | 3 - 4 | 1042 | 80 | ||
| XM_017003877.1 | XP_016859366.1 | 3 - 4 | 340 | 80 | ||
More Information
Set Membership: |
|

ERICH2
Genomic Map