Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301135.1 | NP_001288064.1 | 3 - 4 | 221 | 81 | |
| XM_017012620.1 | XP_016868109.1 | 3 - 4 | 128 | 81 | ||
| GenBank mRNA | AK298469.1 | - | 3 - 4 | 193 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298469.1, NM_001301135.1, XM_017012620.1 transcripts.
Set Membership: |
|

MTERF1
Genomic Map