Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302272.1 | NP_001289201.1 | 1 - 2 | 112 | 112 | |
| NM_006793.4 | NP_006784.1 | 1 - 2 | 112 | 112 | ||
| GenBank mRNA | HY173455.1 | - | 1 - 2 | 112 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302272.1, NM_006793.4 transcripts.
Set Membership: |
|

PRDX3
Genomic Map