Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302272.1 | NP_001289201.1 | 2 - 3 | 248 | 77 | |
| NM_006793.4 | NP_006784.1 | 2 - 3 | 248 | 77 | ||
| GenBank mRNA | AK130897.1 | - | 2 - 3 | 195 | 77 | |
| AK222659.1 | - | 2 - 3 | 178 | 77 | ||
| AK313169.1 | - | 2 - 3 | 186 | 77 | ||
| BC002685.1 | - | 2 - 3 | 179 | 3607558 | 77 | |
| BC007062.1 | - | 2 - 3 | 203 | 3960644 | 77 | |
| BC008435.1 | - | 2 - 3 | 187 | 4081846 | 77 | |
| BC009601.1 | - | 2 - 3 | 180 | 4134666 | 77 | |
| BC021691.1 | - | 2 - 3 | 220 | 3961379 | 77 | |
| BC022373.1 | - | 2 - 3 | 193 | 4775243 | 77 | |
| BC059169.1 | - | 2 - 3 | 180 | 4133576 | 77 | |
| BC111397.1 | - | 2 - 3 | 187 | 4246143 | 77 | |
| BG282768.1 | - | 2 - 3 | 166 | 77 | ||
| BT020007.1 | - | 2 - 3 | 173 | 77 | ||
| BX382389.2 | - | 2 - 3 | 197 | 77 | ||
| CD687044.1 | - | 2 - 3 | 219 | 77 | ||
| CR450344.1 | - | 2 - 3 | 173 | 77 | ||
| D49396.1 | - | 2 - 3 | 179 | 77 | ||
| EF036492.1 | - | 1 - 2 | 148 | 77 | ||
| HY173455.1 | - | 2 - 3 | 248 | 77 | ||
More Information
Set Membership: |
|

PRDX3
Genomic Map