Assay Details
| Gene Symbol: SLC22A18AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302862.1 | NP_001289791.1 | 2 - 3 | 612 | 73 | |
| GenBank mRNA | BC030237.1 | - | 2 - 3 | 168 | 4185944 | 73 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030237.1, NM_001302862.1 transcripts.
Set Membership: |
|

SLC22A18AS
Genomic Map