Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305455.1 | NP_001292384.1 | 1 - 2 | 137 | 92 | |
| GenBank mRNA | AK300573.1 | - | 1 - 2 | 129 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300573.1, NM_001305455.1 transcripts.
Set Membership: |
|

MED20
Genomic Map