Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305456.1 | NP_001292385.1 | 3 - 4 | 417 | 77 | |
| GenBank mRNA | AK097385.1 | - | 3 - 4 | 417 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK097385.1, NM_001305456.1 transcripts.
Set Membership: |
|

MED20
Genomic Map