Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305557.1 | NP_001292486.1 | 1 - 2 | 325 | 84 | |
| GenBank mRNA | BM922767.1 | - | 1 - 2 | 326 | 84 | |
More Information
Set Membership: |
|

TXNL4A
Genomic Map