Assay Details
| Gene Symbol: DLEU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306135.1 | NP_001293064.1 | 1 - 2 | 746 | 81 | |
| GenBank mRNA | AK303567.1 | - | 2 - 3 | 563 | 81 | |
| AY357595.1 | - | 1 - 2 | 503 | 81 | ||
| DN992279.1 | - | 1 - 2 | 329 | 81 | ||
More Information
Set Membership: |
|

DLEU7
Genomic Map