Assay Details
| Gene Symbol: SHH | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310462.1 | NP_001297391.1 | 2 - 3 | 206 | 74 | ||
| NR_132318.1 | 2 - 3 | 206 | 74 | ||||
| NR_132319.1 | 2 - 3 | 206 | 74 | ||||
| XM_011516479.2 | XP_011514781.1 | 2 - 3 | 2948 | 74 | |||
| XM_011516480.2 | XP_011514782.1 | 3 - 4 | 3246 | 74 | |||
| GenBank mRNA | AY927450.1 | - | LIT3478 AY927450 |
2 - 3 | 198 | 74 | |
| AY927452.1 | - | LIT3480 AY927452 |
2 - 3 | 206 | 74 | ||
| AY927453.1 | - | 2 - 3 | 206 | 74 | |||
| AY927454.1 | - | 2 - 3 | 206 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001310462.1, XM_011516479.2, XM_011516480.2 transcripts.
Set Membership: |
|

SHH
Genomic Map