Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012763.1 | NP_001012781.1 | 1 - 2 | 2273 | 102 | |
| GenBank mRNA | Z81148.1 | - | 1 - 2 | 522 | 102 | |
More Information
Set Membership: |
|

GNRHR
Genomic Map