Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317343.1 | NP_001304272.1 | 2 - 3 | 573 | 105 | |
| GenBank mRNA | AK096180.1 | - | 2 - 3 | 440 | 105 | |
| BX538162.1 | - | 1 - 2 | 77 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096180.1 transcripts.
Set Membership: |
|

CDKN2AIP
Genomic Map