Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320395.1 | NP_001307324.1 | 1 - 2 | 192 | 108 | |
| NM_001320397.1 | NP_001307326.1 | 1 - 2 | 192 | 108 | ||
| NM_001320398.1 | NP_001307327.1 | 1 - 2 | 192 | 108 | ||
| NM_001320400.1 | NP_001307329.1 | 1 - 2 | 192 | 108 | ||
| NM_001320401.1 | NP_001307330.1 | 1 - 2 | 192 | 108 | ||
| NM_020357.2 | NP_065090.1 | 1 - 2 | 192 | 108 | ||
| XM_017006922.1 | XP_016862411.1 | 1 - 2 | 106 | 108 | ||
| GenBank mRNA | AB037675.1 | - | 1 - 2 | 88 | 108 | |
| AK098760.1 | - | 1 - 2 | 143 | 108 | ||
| AK222978.1 | - | 1 - 2 | 83 | 108 | ||
| AK314629.1 | - | 1 - 2 | 73 | 108 | ||
| BC013916.1 | - | 1 - 2 | 100 | 3929908 | 108 | |
| BC022001.2 | - | 1 - 2 | 90 | 4823990 | 108 | |
| BF541732.1 | - | 1 - 2 | 91 | 108 | ||
| CD696085.1 | - | 1 - 2 | 107 | 108 | ||
| DA710370.1 | - | 1 - 2 | 82 | 108 | ||
| DB060329.1 | - | 1 - 2 | 103 | 108 | ||
More Information
Set Membership: |
|

PCNP
Genomic Map