Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320397.1 | NP_001307326.1 | 3 - 4 | 380 | 74 | |
| GenBank mRNA | DB060329.1 | - | 3 - 4 | 291 | 74 | |
More Information
Set Membership: |
|

PCNP
Genomic Map