Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320399.1 | NP_001307328.1 | 2 - 3 | 364 | 69 | |
| GenBank mRNA | BI596913.1 | - | 2 - 3 | 290 | 69 | |
More Information
Set Membership: |
|

PCNP
Genomic Map